missing translation for 'onlineSavingsMsg'
Learn More

Sodium Alginate, FCC, 90.8-106%, Spectrum™ Chemical

Catalog No. p-7158176
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606284 2.5 kg Poly Pail
18606285 12 kg Poly Pail
18606286 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606284 Supplier Spectrum Chemical Mfg Cor Supplier No. SO1062.5KG
Only null left

SO106, 9005-38-3, (C6H7O6Na)n

Spectrum™ Chemical Sodium Alginate, FCC is a gum that has no discernible flavor and is used in the food industry as an emulsifier and to increase viscosity. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9005-38-3
Molecular Formula (C6H7O7)A(C6H7O7)BNa
SMILES [Na+].O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C([O-])=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C([O-])=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 90.8 to 106%
Grade FCC
Loss on Drying 0.15
Packaging Poly Pail