missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acid Pyrophosphate, FCC, 93-100.5%, Spectrum™ Chemical

Catalog No. p-7158175
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606278 500 g Poly Bottle
18606279 2.5 kg Poly Pail
18606280 12 kg Poly Pail
18606281 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606278 Supplier Spectrum Chemical Mfg Cor Supplier No. S1099500GM
Only null left

S1099, 7758-16-9, Na2H2P2O7

Spectrum™ Chemical Sodium Acid Pyrophosphate, FCC is leavening agent found in baking powders. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7758-16-9
Molecular Formula Na4O7P2
Molecular Weight (g/mol) 265.90
InChI Key FQENQNTWSFEDLI-UHFFFAOYSA-J
IUPAC Name tetrasodium (phosphonatooxy)phosphonate
SMILES [Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 93 to 100.5%
Grade FCC
Packaging Poly Bottle