missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium acetate trihydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5%
Supplier: Honeywell-Fluka 323186X1KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Sodium Acetate Trihydrate | |
| 6131-90-4 | |
| C2H9NaO5 | |
| MFCD00071557 | |
| sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn | |
| CC(=O)[O-].O.O.O.[Na+] | |
| 136.079 | |
| CHEBI:32138 | |
| ≥99.5% | |
| 1112°F |
| Reag. Ph. Eur. reag. ISO | |
| 6 x 1 kg | |
| CH3COONa · 3H2O | |
| NONH for all modes of transport | |
| AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
| sodium;acetate;trihydrate | |
| 23665404 | |
| 136.08 | |
| ACS/puriss. p.a. |
Chemical Identifiers
| 6131-90-4 | |
| 136.079 | |
| sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn | |
| CHEBI:32138 | |
| CC(=O)[O-].O.O.O.[Na+] |
| C2H9NaO5 | |
| AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
| 23665404 | |
| sodium;acetate;trihydrate |
Safety and Handling
EINECSNumber : 204-823-8
RTECSNumber : AJ4580000