missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate trihydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™

Catalog No. p-7072069
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 1 kg
6 x 2.5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002423 500 g
6002421 1 kg
6002422 2.5 kg
60046129 6 x 1 kg
60046130 6 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6002423 Supplier Honeywell Chemicals Supplier No. 32318500G
Only null left

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

Quantity 500 g
Linear Formula CH3COONa · 3H2O
MDL Number MFCD00071557
UN Number NONH for all modes of transport
Beilstein 3732037
Formula Weight 136.08g/mol
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Ignition Point 1112°F
Chemical Name or Material Sodium Acetate Trihydrate, Reag. Ph. Eur. reag. ISO
EINECSNumber 204-823-8
RTECSNumber AJ4580000