missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate Trihydrate (HPLC), Fisher Chemical

Supplier:  Fisher Chemical FLS2201

 View more versions of this product

Catalog No. S220-1



Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Acetate Trihydrate
6131-90-4
8 to 9.5
1 kg
99 to 101 %
CH3COONa·3H2O
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
CC(=O)[O-].O.O.O.[Na+]
136.079
CHEBI:32138
99 to 101%
Pass Test
1.45g/cm³
58°C
White
Solid
Also meets ACS specifications
C2H9NaO5
MFCD00071557
AYRVGWHSXIMRAB-UHFFFAOYSA-M
sodium;acetate;trihydrate
23665404
136.08
HPLC
Poly Bottle

Chemical Identifiers

6131-90-4
136.079
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
CHEBI:32138
CC(=O)[O-].O.O.O.[Na+]
C2H9NaO5
AYRVGWHSXIMRAB-UHFFFAOYSA-M
23665404
sodium;acetate;trihydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:1
Instability:1

SDS Documents

Documents

Promotions

Promotions