missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 6131-90-4 |
|---|---|
| Molecular Formula | C2H9NaO5 |
| Molecular Weight (g/mol) | 136.079 |
| InChI Key | AYRVGWHSXIMRAB-UHFFFAOYSA-M |
| Synonym | sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn |
| PubChem CID | 23665404 |
| ChEBI | CHEBI:32138 |
| IUPAC Name | sodium;acetate;trihydrate |
| SMILES | CC(=O)[O-].O.O.O.[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
S220-1
|
Fisher Chemical
FLS2201 |
1 kg | Poly Bottle |
Each for $605.00
Case of 6 Each for $3,630.00
|
|
||||
Chemical Identifiers
| 6131-90-4 | |
| 136.079 | |
| sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn | |
| CHEBI:32138 | |
| CC(=O)[O-].O.O.O.[Na+] |
| C2H9NaO5 | |
| AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
| 23665404 | |
| sodium;acetate;trihydrate |
Specifications
| 58°C | |
| White | |
| Solid | |
| 99 to 101 % | |
| CH3COONa·3H2O | |
| sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn | |
| CC(=O)[O-].O.O.O.[Na+] | |
| 136.079 | |
| CHEBI:32138 | |
| 99 to 101% | |
| Pass Test | |
| 1.45g/cm³ |
| 6131-90-4 | |
| 8 to 9.5 | |
| 1 kg | |
| C2H9NaO5 | |
| MFCD00071557 | |
| AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
| sodium;acetate;trihydrate | |
| 23665404 | |
| 136.08 | |
| Also meets ACS specifications | |
| Poly Bottle | |
| Sodium Acetate Trihydrate |
Safety and Handling
CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:1
Flammability:1
Instability:1