missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Acetate, Trihydrate, Granular, FCC, 99-101%, Spectrum™ Chemical
S1114, 6131-90-4, NaC2H3O2·3H2O
$489.18 - $5569.36
Chemical Identifiers
| CAS | 6131-90-4 |
|---|---|
| Molecular Formula | C2H9NaO5 |
| Molecular Weight (g/mol) | 136.08 |
| InChI Key | AYRVGWHSXIMRAB-UHFFFAOYSA-M |
| IUPAC Name | sodium acetate trihydrate |
| SMILES | O.O.O.[Na+].CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606269
|
Spectrum Chemical Mfg Cor
S1114500GM |
500 g | Poly Bottle |
Each for $489.18
|
|
||||
|
18606270
|
Spectrum Chemical Mfg Cor
S111425KG |
2.5 kg | Poly Pail |
Each for $1,456.76
|
|
||||
|
18606271
|
Spectrum Chemical Mfg Cor
S111412KG |
12 kg | Poly Pail |
Each for $3,217.33
|
|
||||
|
18606272
|
Spectrum Chemical Mfg Cor
S111450KGBL |
50 kg | Fiber Drum |
Each for $5,569.36
|
|
||||