missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate, Trihydrate, Granular, FCC, 99-101%, Spectrum™ Chemical

S1114, 6131-90-4, NaC2H3O2·3H2O

Supplier:  Spectrum Chemical Mfg Cor S111412KG

 View more versions of this product

Catalog No. 18606271


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Sodium Acetate, Trihydrate, Granular, FCC is used as food flavorant. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

6131-90-4
36-41%
C2H9NaO5
AYRVGWHSXIMRAB-UHFFFAOYSA-M
sodium acetate trihydrate
99.0 to 101%
1
Poly Pail
12 kg
O.O.O.[Na+].CC([O-])=O
136.08
FCC

Chemical Identifiers

6131-90-4
136.08
sodium acetate trihydrate
C2H9NaO5
AYRVGWHSXIMRAB-UHFFFAOYSA-M
O.O.O.[Na+].CC([O-])=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions