missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate hydrate, Puratronic™, 99.9985% (metals basis)

Catalog No. AA4168522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4168522 100 g
AA4168506 5 g
AA4168514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4168522 Supplier Thermo Scientific Chemicals Supplier No. 04168522

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 41484-91-7 | C2H3NaO2 | 82.03 g/mol

Sodium acetate hydrate is used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 41484-91-7
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00071557
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate hydrate, sodium acetate monohydrate, acetic acid, sodium salt, monohydrate, c2h3o2.na.h2o, naoac water, water sodium acetate, naoac h2o, naoac.h2o, acmc-20ajp8, sodium hydroxide acetic acid
PubChem CID 23681166
SMILES [Na+].CC([O-])=O

Specifications

Melting Point 58°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.9985%
Solubility Information Soluble in water.(726g/L)at 0 C.
IUPAC Name sodium;acetate;hydrate
Formula Weight 82.03 (Anhydrous)
Odor Odorless
Appearance White crystals
Density 1.45
Chemical Name or Material Sodium acetate hydrate
Show More Show Less
EINECSNumber 204-823-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only