CAS RN 41484-91-7
IUPAC Name:
sodium acetate
Synonyms:
sodium acetate|anhydrous sodium acetate|sodium acetate anhydrous|acetic acid, sodium salt|natriumazetat
Molecular Weight (g/mol):
82.03
Molecular Formula:
C2H3NaO2
InChi Key:
VMHLLURERBWHNL-UHFFFAOYSA-M
SMILES:
[Na+].CC([O-])=O
1
–
1
of
1
results
1
Sodium acetate hydrate, Puratronic™, 99.9985% (metals basis)
CAS: 41484-91-7 Molecular Formula: C2H3NaO2 Molecular Weight (g/mol): 82.03 MDL Number: MFCD00071557 InChI Key: VMHLLURERBWHNL-UHFFFAOYSA-M Synonym: sodium acetate hydrate,sodium acetate monohydrate,acetic acid, sodium salt, monohydrate,c2h3o2.na.h2o,naoac water,water sodium acetate,naoac h2o,naoac.h2o,acmc-20ajp8,sodium hydroxide acetic acid PubChem CID: 23681166 SMILES: [Na+].CC([O-])=O