missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium acetate, 3M aq. soln., pH 7.0, autoclaved, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J60899AK
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Sodium acetate | |
| 127-09-3 | |
| NaOOCCH3 | |
| 3M aq. soln. | |
| VMHLLURERBWHNL-UHFFFAOYSA-M | |
| sodium;acetate | |
| 517045 | |
| 82.03 | |
| 7.0 | |
| Liquid |
| autoclaved | |
| C2H3NaO2 | |
| MFCD00012459 | |
| sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
| [Na+].CC([O-])=O | |
| 82.03 | |
| CHEBI:32954 | |
| Odorless | |
| 250 mL |
Chemical Identifiers
| 127-09-3 | |
| 82.03 | |
| VMHLLURERBWHNL-UHFFFAOYSA-M | |
| 517045 | |
| sodium;acetate |
| C2H3NaO2 | |
| MFCD00012459 | |
| sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
| CHEBI:32954 | |
| [Na+].CC([O-])=O |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only