missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate, 3M aq. soln., pH 7.0, autoclaved, Thermo Scientific Chemicals

Catalog No. AAJ60899EQE
Change view
Click to view available options
Quantity:
125 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60899EQE 125 mL
AAJ60899AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60899EQE Supplier Thermo Scientific Chemicals Supplier No. J60899EQE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 127-09-3 | Molecular Formula: C2H3NaO2 | Molecular Weight: 82.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
IUPAC Name sodium;acetate
SMILES [Na+].CC([O-])=O

Specifications

Concentration 3M aq. soln.
Odor Odorless
pH 7.0
Linear Formula NaOOCCH3
Quantity 125 mL
Physical Form Liquid
Formula Weight 82.03
Chemical Name or Material Sodium acetate, autoclaved
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only