Learn More
Sodium acetate, 3M aq. soln., pH 5.2, autoclaved
Description
Sodium acetate buffer solution pH, 5.2 is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as Chip-on-Chip assay and in vitro enzyme inhibition assay.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Concentration | 3M aq. soln. |
| Odor | Odorless |
| Linear Formula | NaOOCCH3 |
| pH | 5.2 |
| Quantity | 125 mL |
| Physical Form | Liquid |
| Solubility Information | Soluble in water (246g/L) at 20°C,and ethanol. |
| Formula Weight | 82.03 |
| Chemical Name or Material | Sodium acetate, Autoclaved |
Safety and Handling
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only