missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate, 3M aq. soln., pH 4.5, autoclaved

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol

Supplier:  Thermo Scientific Chemicals J61288AK

 View more versions of this product

Catalog No. AAJ61288AK


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium acetate
127-09-3
NaOOCCH3
3M aq. soln.
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate
517045
82.03
4.5
Liquid
autoclaved
C2H3NaO2
MFCD00012459
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
[Na+].CC([O-])=O
82.03
CHEBI:32954
Odorless
250 mL

Chemical Identifiers

127-09-3
82.03
VMHLLURERBWHNL-UHFFFAOYSA-M
517045
[Na+].CC([O-])=O
C2H3NaO2
MFCD00012459
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
CHEBI:32954
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only