missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biochrom Sodium Accelerated Buffer 3
Description
Biochrom buffers are guaranteed to give accurate and reproducible results for up to 3 years from manufacture, provided the storage conditions are adhered to. All reagents have stable chemistry and have been formulated to be stored at ambient temperature so, conveniently, do not need refrigeration.
Specifications
Specifications
| Product Type | Buffer |
| Content And Storage | Store at ambient temperature |
| Quantity | 2 L |
| Form | Liquid |