missing translation for 'onlineSavingsMsg'
Learn More

Sodium 9,10-Dimethoxyanthracene-2-sulfonate 98.0+%, TCI America™

Catalog No. p-7118291
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A57011G 1 g
A57015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A57011G Supplier TCI America Supplier No. A57011G
Only null left

Chemical Identifiers

CAS 67580-39-6
Molecular Formula C16H13NaO5S
Molecular Weight (g/mol) 340.325
MDL Number MFCD00042061
InChI Key CBGQZXMEEWCJQD-UHFFFAOYSA-M
Synonym 9,10-Dimethoxyanthracene-2-sulfonic Acid Sodium Salt, IPA-DAS
PubChem CID 4145994
IUPAC Name sodium;9,10-dimethoxyanthracene-2-sulfonate
SMILES COC1=C2C=CC(=CC2=C(C3=CC=CC=C31)OC)S(=O)(=O)[O-].[Na+]

Specifications

Quantity 1 g
Formula Weight 340.32
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 9,10-Dimethoxyanthracene-2-sulfonate [Fluorimetric Ion-Pair Reagent for Amines]