missing translation for 'onlineSavingsMsg'
Learn More

Sodium 7-Amino-2-naphthalenesulfonate 70.0+%, TCI America™

Catalog No. A035525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A035525G 25 g
A0355500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A035525G Supplier TCI America Supplier No. A035525G

Chemical Identifiers

CAS 5412-82-8
Molecular Formula C10H8NNaO3S
Molecular Weight (g/mol) 245.228
MDL Number MFCD00053436
InChI Key BFGOSNKRBCAENG-UHFFFAOYSA-M
PubChem CID 23678693
IUPAC Name sodium;7-aminonaphthalene-2-sulfonate
SMILES C1=CC(=CC2=C1C=CC(=C2)S(=O)(=O)[O-])N.[Na+]

Specifications

Quantity 25 g
Synonym Cassella′Cs Acid F Sodium Salt, 2-Naphthylamine-7-sulfonic Acid Sodium Salt, 7-Amino-2-naphthalenesulfonic Acid Sodium Salt
Formula Weight 245.23
Percent Purity ≥70.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 7-Amino-2-naphthalenesulfonate