missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 4-Hydroxybenzoate 99.0+%, TCI America™
Supplier: TCI America H0221500G
Specifications
| Sodium 4-Hydroxybenzoate | |
| 300°C | |
| C7H5NaO3 | |
| 500 g | |
| ZLVSYODPTJZFMK-UHFFFAOYSA-M | |
| sodium 4-hydroxybenzoate | |
| 16219477 | |
| ≥99.0% (T) |
| 114-63-6 | |
| White | |
| MFCD00016530 | |
| sodium 4-hydroxybenzoate, sodium p-hydroxybenzoate, 4-hydroxybenzoic acid sodium salt, sodium paraben, benzoic acid, 4-hydroxy-, monosodium salt, p-hydroxybenzoic acid sodium salt, monosodium p-hydroxybenzoate, monosodium 4-hydroxybenzoate, benzoic acid, p-hydroxy-, sodium salt, benzoic acid, 4-hydroxy-, sodium salt 1:1 | |
| [Na+].OC1=CC=C(C=C1)C([O-])=O | |
| 160.10 | |
| 160.10 | |
| Crystalline Powder |
Chemical Identifiers
| 114-63-6 | |
| 160.10 | |
| ZLVSYODPTJZFMK-UHFFFAOYSA-M | |
| 16219477 | |
| [Na+].OC1=CC=C(C=C1)C([O-])=O |
| C7H5NaO3 | |
| MFCD00016530 | |
| sodium 4-hydroxybenzoate, sodium p-hydroxybenzoate, 4-hydroxybenzoic acid sodium salt, sodium paraben, benzoic acid, 4-hydroxy-, monosodium salt, p-hydroxybenzoic acid sodium salt, monosodium p-hydroxybenzoate, monosodium 4-hydroxybenzoate, benzoic acid, p-hydroxy-, sodium salt, benzoic acid, 4-hydroxy-, sodium salt 1:1 | |
| sodium 4-hydroxybenzoate |
Safety and Handling
EINECSNumber : (3)-1585&(9)-1094
RTECSNumber : DH2900000
TSCA : Yes