missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-Hydroxybenzoate 99.0+%, TCI America™

Catalog No. p-7130419
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H022125G 25 g
H0221500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H022125G Supplier TCI America Supplier No. H022125G

Chemical Identifiers

CAS 114-63-6
Molecular Formula C7H5NaO3
Molecular Weight (g/mol) 160.10
MDL Number MFCD00016530
InChI Key ZLVSYODPTJZFMK-UHFFFAOYSA-M
Synonym sodium 4-hydroxybenzoate, sodium p-hydroxybenzoate, 4-hydroxybenzoic acid sodium salt, sodium paraben, benzoic acid, 4-hydroxy-, monosodium salt, p-hydroxybenzoic acid sodium salt, monosodium p-hydroxybenzoate, monosodium 4-hydroxybenzoate, benzoic acid, p-hydroxy-, sodium salt, benzoic acid, 4-hydroxy-, sodium salt 1:1
PubChem CID 16219477
IUPAC Name sodium 4-hydroxybenzoate
SMILES [Na+].OC1=CC=C(C=C1)C([O-])=O

Specifications

Melting Point 300°C
Color White
Quantity 25 g
Formula Weight 160.10
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 4-Hydroxybenzoate
EINECSNumber (3)-1585&(9)-1094
RTECSNumber DH2900000
TSCA Yes