missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 4-Hydroxybenzenesulfonate 98.0+%, TCI America™
Supplier: TCI America P010525G
Specifications
| Sodium 4-Hydroxybenzenesulfonate | |
| White-Yellow | |
| MFCD00044734 | |
| sodium 4-hydroxybenzenesulfonate, monosodium 4-phenolsulfonate, sodium p-hydroxybenzenesulfonate, p-phenolsulfonic acid sodium salt, unii-95h8x8yvqr, benzenesulfonic acid, 4-hydroxy-, monosodium salt, sodium p-phenolsulfonate, 95h8x8yvqr, 4-phenolsulfonic acid, sodium salt, sodium 4-hydroxybenzene-1-sulfonate | |
| C1=CC(=CC=C1O)S(=O)(=O)[O-].[Na+] | |
| 196.152 | |
| 196.15 | |
| Crystalline Powder |
| 825-90-1 | |
| C6H5NaO4S | |
| 25 g | |
| BYMHXIQVEAYSJD-UHFFFAOYSA-M | |
| sodium;4-hydroxybenzenesulfonate | |
| 4379756 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 825-90-1 | |
| 196.152 | |
| BYMHXIQVEAYSJD-UHFFFAOYSA-M | |
| 4379756 | |
| C1=CC(=CC=C1O)S(=O)(=O)[O-].[Na+] |
| C6H5NaO4S | |
| MFCD00044734 | |
| sodium 4-hydroxybenzenesulfonate, monosodium 4-phenolsulfonate, sodium p-hydroxybenzenesulfonate, p-phenolsulfonic acid sodium salt, unii-95h8x8yvqr, benzenesulfonic acid, 4-hydroxy-, monosodium salt, sodium p-phenolsulfonate, 95h8x8yvqr, 4-phenolsulfonic acid, sodium salt, sodium 4-hydroxybenzene-1-sulfonate | |
| sodium;4-hydroxybenzenesulfonate |
Safety and Handling
EINECSNumber : (3)-1958
TSCA : Yes