missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 4-Hydroxybenzenesulfonate 98.0+%, TCI America™
$41.02 - $415.68
Chemical Identifiers
| CAS | 825-90-1 |
|---|---|
| Molecular Formula | C6H5NaO4S |
| Molecular Weight (g/mol) | 196.152 |
| MDL Number | MFCD00044734 |
| InChI Key | BYMHXIQVEAYSJD-UHFFFAOYSA-M |
| Synonym | sodium 4-hydroxybenzenesulfonate, monosodium 4-phenolsulfonate, sodium p-hydroxybenzenesulfonate, p-phenolsulfonic acid sodium salt, unii-95h8x8yvqr, benzenesulfonic acid, 4-hydroxy-, monosodium salt, sodium p-phenolsulfonate, 95h8x8yvqr, 4-phenolsulfonic acid, sodium salt, sodium 4-hydroxybenzene-1-sulfonate |
| PubChem CID | 4379756 |
| IUPAC Name | sodium;4-hydroxybenzenesulfonate |
| SMILES | C1=CC(=CC=C1O)S(=O)(=O)[O-].[Na+] |
Chemical Identifiers
| 825-90-1 | |
| 196.152 | |
| BYMHXIQVEAYSJD-UHFFFAOYSA-M | |
| 4379756 | |
| C1=CC(=CC=C1O)S(=O)(=O)[O-].[Na+] |
| C6H5NaO4S | |
| MFCD00044734 | |
| sodium 4-hydroxybenzenesulfonate, monosodium 4-phenolsulfonate, sodium p-hydroxybenzenesulfonate, p-phenolsulfonic acid sodium salt, unii-95h8x8yvqr, benzenesulfonic acid, 4-hydroxy-, monosodium salt, sodium p-phenolsulfonate, 95h8x8yvqr, 4-phenolsulfonic acid, sodium salt, sodium 4-hydroxybenzene-1-sulfonate | |
| sodium;4-hydroxybenzenesulfonate |
Specifications
| 825-90-1 | |
| C6H5NaO4S | |
| 25 g | |
| BYMHXIQVEAYSJD-UHFFFAOYSA-M | |
| sodium;4-hydroxybenzenesulfonate | |
| 4379756 | |
| ≥98.0% (HPLC,T) | |
| Sodium 4-Hydroxybenzenesulfonate |
| White-Yellow | |
| MFCD00044734 | |
| sodium 4-hydroxybenzenesulfonate, monosodium 4-phenolsulfonate, sodium p-hydroxybenzenesulfonate, p-phenolsulfonic acid sodium salt, unii-95h8x8yvqr, benzenesulfonic acid, 4-hydroxy-, monosodium salt, sodium p-phenolsulfonate, 95h8x8yvqr, 4-phenolsulfonic acid, sodium salt, sodium 4-hydroxybenzene-1-sulfonate | |
| C1=CC(=CC=C1O)S(=O)(=O)[O-].[Na+] | |
| 196.152 | |
| 196.15 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1958
TSCA : Yes