missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-Hydroxyazobenzene-4'-sulfonate Hydrate 98.0+%, TCI America™

Catalog No. H03275G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H03275G 5 g
H032725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H03275G Supplier TCI America Supplier No. H03275G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2623-36-1
Molecular Formula C12H9N2NaO4S
Molecular Weight (g/mol) 300.264
MDL Number MFCD00068861
InChI Key QZIOKUAQNKVSBO-UHFFFAOYSA-M
PubChem CID 101973309
IUPAC Name sodium;4-[(4-sulfophenyl)diazenyl]phenolate
SMILES C1=CC(=CC=C1N=NC2=CC=C(C=C2)S(=O)(=O)O)[O-].[Na+]

Specifications

Quantity 5 g
Synonym 4-Hydroxyazobenzene-4′C-sulfonic Acid Sodium Salt
Formula Weight 300.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 4-Hydroxyazobenzene-4′-sulfonate Hydrate