accessibility menu, dialog, popup

UserName

CAS RN 2623-36-1

IUPAC Name: sodium 4-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]benzene-1-sulfonate
Synonyms: sodium 4-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]benzenesulfonate
Molecular Weight (g/mol): 300.26
Molecular Formula: C12H9N2NaO4S
InChi Key: SYZKFRYUZILKHZ-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC=C(NN=C2C=CC(=O)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results