missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-Ethylbenzenesulfonate 98.0+%, TCI America™

Catalog No. E0104500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0104500G 500 g
E010425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0104500G Supplier TCI America Supplier No. E0104500G

Chemical Identifiers

CAS 14995-38-1
Molecular Formula C8H9NaO3S
Molecular Weight (g/mol) 208.21
MDL Number MFCD00070568
InChI Key LLELGQVCQVOGMA-UHFFFAOYSA-M
Synonym 4-Ethylbenzenesulfonic Acid Sodium Salt
PubChem CID 23675352
IUPAC Name sodium 4-ethylbenzene-1-sulfonate
SMILES [Na+].CCC1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Color White
Quantity 500 g
Formula Weight 208.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 4-Ethylbenzenesulfonate
EINECSNumber (3)-3127
TSCA Yes