accessibility menu, dialog, popup

UserName

CAS RN 14995-38-1

NaH3CSOOO

CAS RN 14995-38-1

IUPAC Name: sodium 4-ethylbenzene-1-sulfonate
Synonyms: sodium 4-ethylbenzenesulfonate
Molecular Weight (g/mol): 208.21
Molecular Formula: C8H9NaO3S
InChi Key: LLELGQVCQVOGMA-UHFFFAOYSA-M
SMILES: [Na+].CCC1=CC=C(C=C1)S([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results