missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-Aminosalicylate Dihydrate 98.0+%, TCI America™

Catalog No. A042325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A042325G 25 g
A0423500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A042325G Supplier TCI America Supplier No. A042325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6018-19-5
Molecular Formula C7H10NNaO5
Molecular Weight (g/mol) 211.149
MDL Number MFCD00064392
InChI Key GMUQJDAYXZXBOT-UHFFFAOYSA-M
Synonym sodium 4-aminosalicylate dihydrate, sodium 4-amino-2-hydroxybenzoate dihydrate, parasal sodium, 4-aminosalicylic acid sodium salt dihydrate, unii-s38b9w6axw, aminosalicylate sodium, sodium aminosalicylate, sodium p.a.s., s38b9w6axw, aminosalicylate sodium usp
PubChem CID 16211148
IUPAC Name sodium;4-amino-2-hydroxybenzoate;dihydrate
SMILES C1=CC(=C(C=C1N)O)C(=O)[O-].O.O.[Na+]

Specifications

Quantity 25 g
Formula Weight 175.12
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 4-Aminosalicylate Dihydrate
RTECSNumber VO1700000
TSCA Yes