missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-aminosalicylate dihydrate, 98%

Catalog No. AAJ6444936
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6444936 500 g
AAJ6444914 25 g
AAJ6444922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6444936 Supplier Thermo Scientific Chemicals Supplier No. J6444936

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6018-19-5 | C7H10NNaO5 | 211.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6018-19-5
Molecular Formula C7H10NNaO5
Molecular Weight (g/mol) 211.149
MDL Number MFCD00151044
InChI Key GMUQJDAYXZXBOT-UHFFFAOYSA-M
Synonym sodium 4-aminosalicylate dihydrate, sodium 4-amino-2-hydroxybenzoate dihydrate, parasal sodium, 4-aminosalicylic acid sodium salt dihydrate, unii-s38b9w6axw, aminosalicylate sodium, sodium aminosalicylate, sodium p.a.s., s38b9w6axw, aminosalicylate sodium usp
PubChem CID 16211148
IUPAC Name sodium;4-amino-2-hydroxybenzoate;dihydrate
SMILES C1=CC(=C(C=C1N)O)C(=O)[O-].O.O.[Na+]

Specifications

Melting Point 250°C
Color White
Quantity 500 g
Sensitivity Air and light sensitive
Formula Weight 211.14 (175.12ahy)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sodium 4-aminosalicylate dihydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-091-2
RTECSNumber VO1700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only