missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-Amino-5-hydroxy-2,7-naphthalenedisulfonate Hydrate 80.0+%, TCI America™

Catalog No. a0362500g
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0362500G 500 g
A036225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0362500G Supplier TCI America Supplier No. A0362500G
Only null left

Chemical Identifiers

CAS 5460-09-3
Molecular Formula C10H8NNaO7S2
Molecular Weight (g/mol) 341.28
MDL Number MFCD00036444
InChI Key QPILZZVXGUNELN-UHFFFAOYSA-M

Specifications

Quantity 500 g
Synonym monosodium 8-amino-1-naphthol-3,6-disulfonate monohydrate, sodium 4-amino-5-hydroxy-7-sulfonaphthalene-2-sulfonate hydrate, 8-amino-1-naphthol-3,6-disulfonic acid monosodium salt, 4-amino-5-hydroxynaphthalene-2,7-disulfonic acid monosodium salt hydrate, dsstox_cid_26980, dsstox_rid_82035, dsstox_gsid_46980
SMILES [Na+].NC1=C2C(O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S([O-])(=O)=O
IUPAC Name sodium 5-amino-4-hydroxy-7-sulfonaphthalene-2-sulfonate
PubChem CID 60196419
Formula Weight 341.28
Percent Purity ≥80.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 4-Amino-5-hydroxy-2,7-naphthalenedisulfonate Hydrate
EINECSNumber (4)-0523
TSCA Yes