missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-Amino-5-hydroxy-2,7-naphthalenedisulfonate Hydrate 80.0+%, TCI America™

Catalog No. A036225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A036225G 25 g
A0362500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A036225G Supplier TCI America Supplier No. A036225G

Chemical Identifiers

CAS 5460-09-3
Molecular Formula C10H8NNaO7S2
Molecular Weight (g/mol) 341.28
MDL Number MFCD00036444
InChI Key QPILZZVXGUNELN-UHFFFAOYSA-M

Specifications

Quantity 25 g
Synonym h acid sodium, 1-amino-8-naphthol-3,6-disulfonic acid monosodiumsalt, 4-amino-5-hydroxynaphthalene-2,7-disulfonic acid 2-sodium salt
SMILES C1=C2C=C(C=C(C2=C(C=C1S(=O)(=O)O)N)[O-])S(=O)(=O)O.[Na+]
IUPAC Name sodium;8-amino-3,6-disulfonaphthalen-1-olate
PubChem CID 102116466
Formula Weight 341.28
Percent Purity ≥80.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 4-Amino-5-hydroxy-2,7-naphthalenedisulfonate Hydrate
EINECSNumber (4)-0523
TSCA Yes