missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-(Trimethylsilyl)-1-propanesulfonate 98.0+%, TCI America™

$397.05 - $1985.22

Chemical Identifiers

CAS 2039-96-5
Molecular Formula C6H15NaO3SSi
Molecular Weight (g/mol) 218.32
MDL Number MFCD00007537
InChI Key HWEXKRHYVOGVDA-UHFFFAOYSA-M
Synonym sodium 3-trimethylsilyl propane-1-sulfonate, 3-trimethylsilyl-1-propanesulfonic acid sodium salt, dss sodium salt, sodium 3-trimethylsilyl-1-propanesulfonate, 1-propanesulfonic acid, 3-trimethylsilyl-, sodium salt, 2,2-dimethyl-2-silapentane-5-sulfonate sodium salt, sodium 3-sulfopropyltrimethylsilane, 1-propanesulfonic acid, 3-trimethylsilyl-, sodium salt 1:1, sodium 2,2-dimethyl-2-silapentane-5-sulfonate, 3-trimethylsilyl propanesulfonic acid
PubChem CID 5167273
IUPAC Name sodium 3-(trimethylsilyl)propane-1-sulfonate
SMILES [Na+].C[Si](C)(C)CCCS([O-])(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T16381G
View Documents
TCI America
T16381G
1 g
Each for $397.05
Only null left
 
T16385G
View Documents
TCI America
T16385G
5 g
Each for $1,985.22
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2039-96-5
218.32
HWEXKRHYVOGVDA-UHFFFAOYSA-M
5167273
[Na+].C[Si](C)(C)CCCS([O-])(=O)=O
C6H15NaO3SSi
MFCD00007537
sodium 3-trimethylsilyl propane-1-sulfonate, 3-trimethylsilyl-1-propanesulfonic acid sodium salt, dss sodium salt, sodium 3-trimethylsilyl-1-propanesulfonate, 1-propanesulfonic acid, 3-trimethylsilyl-, sodium salt, 2,2-dimethyl-2-silapentane-5-sulfonate sodium salt, sodium 3-sulfopropyltrimethylsilane, 1-propanesulfonic acid, 3-trimethylsilyl-, sodium salt 1:1, sodium 2,2-dimethyl-2-silapentane-5-sulfonate, 3-trimethylsilyl propanesulfonic acid
sodium 3-(trimethylsilyl)propane-1-sulfonate

Specifications

2039-96-5
C6H15NaO3SSi
1 g
HWEXKRHYVOGVDA-UHFFFAOYSA-M
sodium 3-(trimethylsilyl)propane-1-sulfonate
5167273
≥98.0% (T)
Sodium 3-(Trimethylsilyl)-1-propanesulfonate [1H NMR Standard for D2O Solvent]
White
MFCD00007537
sodium 3-trimethylsilyl propane-1-sulfonate, 3-trimethylsilyl-1-propanesulfonic acid sodium salt, dss sodium salt, sodium 3-trimethylsilyl-1-propanesulfonate, 1-propanesulfonic acid, 3-trimethylsilyl-, sodium salt, 2,2-dimethyl-2-silapentane-5-sulfonate sodium salt, sodium 3-sulfopropyltrimethylsilane, 1-propanesulfonic acid, 3-trimethylsilyl-, sodium salt 1:1, sodium 2,2-dimethyl-2-silapentane-5-sulfonate, 3-trimethylsilyl propanesulfonic acid
[Na+].C[Si](C)(C)CCCS([O-])(=O)=O
218.32
218.32
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents