accessibility menu, dialog, popup

UserName

CAS RN 2039-96-5

IUPAC Name: sodium 3-(trimethylsilyl)propane-1-sulfonate
Synonyms: sodium 3-(trimethylsilyl)propane-1-sulfonate
Molecular Weight (g/mol): 218.32
Molecular Formula: C6H15NaO3SSi
InChi Key: HWEXKRHYVOGVDA-UHFFFAOYSA-M
SMILES: [Na+].C[Si](C)(C)CCCS([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results