missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-(Trimethylsilyl)-1-propanesulfonate 98.0+%, TCI America™

Catalog No. T16385G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16385G 5 g
T16381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16385G Supplier TCI America Supplier No. T16385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2039-96-5
Molecular Formula C6H15NaO3SSi
Molecular Weight (g/mol) 218.32
MDL Number MFCD00007537
InChI Key HWEXKRHYVOGVDA-UHFFFAOYSA-M
Synonym sodium 3-trimethylsilyl propane-1-sulfonate, 3-trimethylsilyl-1-propanesulfonic acid sodium salt, dss sodium salt, sodium 3-trimethylsilyl-1-propanesulfonate, 1-propanesulfonic acid, 3-trimethylsilyl-, sodium salt, 2,2-dimethyl-2-silapentane-5-sulfonate sodium salt, sodium 3-sulfopropyltrimethylsilane, 1-propanesulfonic acid, 3-trimethylsilyl-, sodium salt 1:1, sodium 2,2-dimethyl-2-silapentane-5-sulfonate, 3-trimethylsilyl propanesulfonic acid
PubChem CID 5167273
IUPAC Name sodium 3-(trimethylsilyl)propane-1-sulfonate
SMILES [Na+].C[Si](C)(C)CCCS([O-])(=O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 218.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 3-(Trimethylsilyl)-1-propanesulfonate [1H NMR Standard for D2O Solvent]