missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-bromopropanesulfonate, 97%

Catalog No. AAA1993006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1993006 5 g
AAA1993014 25 g
AAA1993022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1993006 Supplier Thermo Scientific Chemicals Supplier No. A1993006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 55788-44-8 | C3H7BrNaO3S | 226.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55788-44-8
Molecular Formula C3H7BrNaO3S
Molecular Weight (g/mol) 226.04
MDL Number MFCD00024999
InChI Key ONPNLMMBTJZJRQ-UHFFFAOYSA-N
Synonym sodium 3-bromopropanesulfonate, sodium 3-bromopropane-1-sulfonate, 3-bromopropanesulfonic acid sodium salt, acmc-209lpd, sodium 3-bromo-1-propanesulfonate, sodium 3-bromo-propane-1-sulphonate, 3-bromopropane sulfonic acid sodium salt, 3-bromo-1-propanesulfonic acid sodium salt, 1-propanesulfonic acid, 3-bromo-, sodium salt
PubChem CID 23679056
IUPAC Name sodium;3-bromopropane-1-sulfonate
SMILES [Na].OS(=O)(=O)CCCBr

Specifications

Melting Point ∼256°C (decomposition)
Quantity 5 g
Formula Weight 225.05
Percent Purity 97%
Chemical Name or Material Sodium 3-bromopropanesulfonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only