missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-(4-Hydroxy-3-methoxyphenyl)lactate 99.0+%, TCI America™

Catalog No. H0538100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0538100MG 100 mg
H05381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0538100MG Supplier TCI America Supplier No. H0538100MG

Chemical Identifiers

CAS 77305-02-3
Molecular Formula C10H11NaO5
Molecular Weight (g/mol) 234.183
MDL Number MFCD00056719
InChI Key VYPSMFXPMZZSSK-UHFFFAOYSA-M
Synonym 3-(4-Hydroxy-3-methoxyphenyl)lactic Acid Sodium Salt, Sodium 2-Hydroxy-3-(4-hydroxy-3-methoxyphenyl)propionate, 2-Hydroxy-3-(4-hydroxy-3-methoxyphenyl)propionic Acid Sodium Salt
PubChem CID 44630328
IUPAC Name sodium;2-hydroxy-3-(4-hydroxy-3-methoxyphenyl)propanoate
SMILES COC1=C(C=CC(=C1)CC(C(=O)[O-])O)O.[Na+]

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 234.18
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 3-(4-Hydroxy-3-methoxyphenyl)lactate