accessibility menu, dialog, popup

UserName

H3COOOOHHONa

CAS RN 77305-02-3

IUPAC Name: sodium 2-hydroxy-3-(4-hydroxy-3-methoxyphenyl)propanoate
Synonyms: sodium 2-hydroxy-3-(4-hydroxy-3-methoxyphenyl)propanoate
Molecular Weight (g/mol): 234.18
Molecular Formula: C10H11NaO5
InChi Key: VYPSMFXPMZZSSK-UHFFFAOYNA-M
SMILES: [Na+].COC1=C(O)C=CC(CC(O)C([O-])=O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results