missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-Naphthol-6-sulfonate Hydrate 97.0+%, TCI America™

Catalog No. N0035500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0035500G 500 g
N003525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0035500G Supplier TCI America Supplier No. N0035500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135-76-2
Molecular Formula C10H7NaO4S
Molecular Weight (g/mol) 246.212
MDL Number MFCD00070488
InChI Key ZPWQALCOMQRMRK-UHFFFAOYSA-M
PubChem CID 23667648
IUPAC Name sodium;6-hydroxynaphthalene-2-sulfonate
SMILES C1=CC2=C(C=CC(=C2)S(=O)(=O)[O-])C=C1O.[Na+]

Specifications

Quantity 500 g
Synonym 6-Hydroxy-2-naphthalenesulfonic Acid Sodium Salt, Schaeffer′s Acid Sodium Salt, Sodium 6-Hydroxy-2-naphthalenesulfonate, 2-Naphthol-6-sulfonic Acid Sodium Salt
Formula Weight 246.21
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2-Naphthol-6-sulfonate Hydrate
EINECSNumber (4)-0479
TSCA Yes