missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-Methyl-2-propene-1-sulfonate 98.0+%, TCI America™

Catalog No. M140825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M140825G 25 g
M1408500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M140825G Supplier TCI America Supplier No. M140825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1561-92-8
Molecular Formula C4H7NaO3S
Molecular Weight (g/mol) 158.15
MDL Number MFCD00065344
InChI Key SZHIIIPPJJXYRY-UHFFFAOYSA-M
Synonym Methallylsulfonic Acid Sodium Salt, Sodium Methallylsulfonate, 2-Methyl-2-propene-1-sulfonic Acid Sodium Salt
PubChem CID 23663624
IUPAC Name sodium 2-methylprop-2-ene-1-sulfonate
SMILES [Na+].CC(=C)CS([O-])(=O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 158.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2-Methyl-2-propene-1-sulfonate
EINECSNumber (2)-1590
TSCA Yes