accessibility menu, dialog, popup

UserName

NaCH3CH2SOOO

CAS RN 1561-92-8

IUPAC Name: sodium 2-methylprop-2-ene-1-sulfonate
Synonyms: sodium 2-methylprop-2-ene-1-sulfonate
Molecular Weight (g/mol): 158.15
Molecular Formula: C4H7NaO3S
InChi Key: SZHIIIPPJJXYRY-UHFFFAOYSA-M
SMILES: [Na+].CC(=C)CS([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results