missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-mercaptoethanesulfonate, Analytical Standard, 98.0% (titration), MilliporeSigma™ Supelco™

Catalog No. 111013440
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013440 5 g
111013438 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013440 Supplier Merck Emd Millipore Supplier No. M15115G
Only null left

Specifications

Packaging Poly Bottle
CAS 19767-45-4
Physical Form Neat
Linear Formula HSCH2CH2SO3Na
MDL Number MFCD00007535
Synonym 2-Mercaptoethanesulfonic acid sodium salt; Coenzyme M sodium salt; HS-CoM Na; MESNA
Quantity 5 g
Formula Weight 164.18
Percent Purity ≥98.0% (titration)
Grade Analytical Standard