missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-mercaptoethanesulfonate, 98.0% (titration), MilliporeSigma™ Supelco™

Catalog No. 111013439
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013439 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013439 Supplier Merck Emd Millipore Supplier No. M151110G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 19767-45-4
Physical Form Neat
Linear Formula HSCH2 CH2 SO3 Na
MDL Number MFCD00007535
Synonym 2-Mercaptoethanesulfonic acid sodium salt; Coenzyme M sodium salt; HS-CoM Na; MESNA
Type Analytical Standard
Quantity 10 g
Formula Weight 164.18
Percent Purity ≥98% (titration)
Grade Analytical Standard