missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-Mercaptobenzothiazole 97.0+%, TCI America™

Catalog No. p-7131957
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M005725G 25 g
M0057500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M005725G Supplier TCI America Supplier No. M005725G
Only null left

Chemical Identifiers

CAS 2492-26-4
Molecular Formula C7H5NNaS2
Molecular Weight (g/mol) 190.234
MDL Number MFCD00065334
InChI Key KRXFTOUYGXMRRU-UHFFFAOYSA-N
Synonym Sodium 2-Benzothiazolethiolate, 2-Mercaptobenzothiazole Sodium Salt
PubChem CID 24195611
IUPAC Name 3H-1,3-benzothiazole-2-thione;sodium
SMILES C1=CC=C2C(=C1)NC(=S)S2.[Na]

Specifications

pH 10-11 (1% aq)
Quantity 25 g
UN Number 3077
Formula Weight 189.23
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2-Mercaptobenzothiazole
EINECSNumber (5)-0243
RTECSNumber DL6725000
TSCA Yes