missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-chloroethanesulfonate hydrate, 98+% (dry wt.), water <10%

Catalog No. AAA1999506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1999506 5 g
AAA1999514 25 g
AAA1999522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1999506 Supplier Thermo Scientific Chemicals Supplier No. A1999506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 15484-44-3 | C2H4ClNaO3S | 166.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15484-44-3
Molecular Formula C2H4ClNaO3S
Molecular Weight (g/mol) 166.55
MDL Number MFCD00149546 MFCD00007531
InChI Key BVIXLMYIFZGRBH-UHFFFAOYSA-M
Synonym sodium 2-chloroethylsulfonate, 2-chloroethane sulfonic acid sodium salt, sodium .beta.-chloroethanesulfonate, sodium 2-chloroethane-1-sulfonic acid, sodium 2-chloroethanesulfonic acid
PubChem CID 54611855
SMILES [Na+].[O-]S(=O)(=O)CCCl

Specifications

Melting Point ∼292°C (decomposition)
Quantity 5 g
Beilstein 5649202
Sensitivity Hygroscopic
IUPAC Name sodium;2-chloroethanesulfonic acid
Formula Weight 166.56 (Anhydrous)
Percent Purity ≥98%
Assay (dry wt.), water <10%
Chemical Name or Material Sodium 2-chloroethanesulfonate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-508-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only