accessibility menu, dialog, popup

UserName

CAS RN 15484-44-3

IUPAC Name: sodium 2-chloroethane-1-sulfonate
Synonyms: sodium 2-chloroethanesulfonate
Molecular Weight (g/mol): 166.55
Molecular Formula: C2H4ClNaO3S
InChi Key: BVIXLMYIFZGRBH-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)CCCl
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)