missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-Chloroethanesulfonate (ca. 13% in Water), TCI America™

Catalog No. C111025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C111025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C111025G Supplier TCI America Supplier No. C111025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15484-44-3
Molecular Formula C2H4ClNaO3S
Molecular Weight (g/mol) 166.55
MDL Number MFCD00149546 MFCD00007531
InChI Key BVIXLMYIFZGRBH-UHFFFAOYSA-M
Synonym sodium 2-chloroethylsulfonate, 2-chloroethane sulfonic acid sodium salt, sodium .beta.-chloroethanesulfonate, sodium 2-chloroethane-1-sulfonic acid, sodium 2-chloroethanesulfonic acid
PubChem CID 54611855
IUPAC Name sodium 2-chloroethane-1-sulfonate
SMILES [Na+].[O-]S(=O)(=O)CCCl

Specifications

Color Yellow
Quantity 25 g
Formula Weight 166.55
Physical Form Liquid
Chemical Name or Material Sodium 2-Chloroethanesulfonate (ca. 13% in Water)