missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-Amino-4-chloro-5-methylbenzenesulfonate 98.0+%, TCI America™

Catalog No. A0896500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0896500G 500 g
A089625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0896500G Supplier TCI America Supplier No. A0896500G
Only null left

Chemical Identifiers

CAS 6627-59-4
Molecular Formula C7H8ClNNaO3S
Molecular Weight (g/mol) 244.645
MDL Number MFCD00070551
InChI Key OSSZFTJQTIEJDM-UHFFFAOYSA-N
Synonym 5-Chloro-4-methylaniline-2-sulfonic Acid Sodium Salt, 5-Chloro-p-toluidine-2-sulfonic Acid Sodium Salt, Sodium 4-Amino-2-chlorotoluene-5-sulfonate, 4-Amino-2-chlorotoluene-5-sulfonic Acid Sodium Salt, Sodium 5-Chloro-4-methylaniline-2-sulfonate, Sodium 5-
PubChem CID 24187155
IUPAC Name 2-amino-4-chloro-5-methylbenzenesulfonic acid;sodium
SMILES CC1=CC(=C(C=C1Cl)N)S(=O)(=O)O.[Na]

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 243.64
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2-Amino-4-chloro-5-methylbenzenesulfonate
EINECSNumber (3)-2024
TSCA Yes