accessibility menu, dialog, popup

UserName

CAS RN 6627-59-4

NaCH3NH2ClSOHOO

CAS RN 6627-59-4

IUPAC Name: 2-amino-4-chloro-5-methylbenzene-1-sulfonic acid sodium
Synonyms: 2-amino-4-chloro-5-methylbenzenesulfonic acid sodium
Molecular Weight (g/mol): 244.64
Molecular Formula: C7H8ClNNaO3S
InChi Key: OSSZFTJQTIEJDM-UHFFFAOYSA-N
SMILES: [Na].CC1=CC(=C(N)C=C1Cl)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results