missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2,4-pentanedionate hydrate, 94%

Catalog No. AAB2400918
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2400918 50 g
AAB2400930 250 g
AAB240090B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2400918 Supplier Thermo Scientific Chemicals Supplier No. B2400918
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 86891-03-4 | C5H7NaO2 | 122.10 g/mol

Sodium 2,4-pentanedionate hydrate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 2,4-pentanedionate hydrate is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are strong oxidizing agents, water/ moisture, acids.

Chemical Identifiers

CAS 86891-03-4
Molecular Formula C5H7NaO2
Molecular Weight (g/mol) 122.10
MDL Number MFCD00078034
InChI Key AIWZOHBYSFSQGV-UHFFFAOYSA-M
Synonym sodium 2,4-pentanedionate, sodium 2e-4-oxopent-2-en-2-olate hydrate
PubChem CID 90476915
IUPAC Name sodium;(E)-4-oxopent-2-en-2-olate;hydrate
SMILES [Na+].CC([O-])=CC(C)=O

Specifications

Melting Point 228°C to 230°C
Quantity 50 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 122.11 (Anhydrous)
Percent Purity 95%
Chemical Name or Material Sodium 2,4-pentanedionate hydrate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 239-448-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only