missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2,4-Dinitrobenzenesulfonate Hydrate 98.0+%, TCI America™

Catalog No. D082125G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D082125G 25 g
D0821500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. D082125G missing translation for 'mfr' TCI America missing translation for 'supplierNo' D082125G

Chemical Identifiers

CAS 885-62-1
Molecular Formula C6H3N2NaO7S
Molecular Weight (g/mol) 270.15
MDL Number MFCD00007471,MFCD10567393
InChI Key GSBYVRKLPCSLNV-UHFFFAOYSA-M
Synonym sodium 2,4-dinitrobenzenesulfonate hydrate, acmc-209quh, sodium dnbs hydrate, 2,4-dinitrobenzenesulfonic acid sodium salt hydrate
PubChem CID 70700115
IUPAC Name sodium 2,4-dinitrobenzene-1-sulfonate
SMILES [Na+].[O-][N+](=O)C1=CC=C(C(=C1)[N+]([O-])=O)S([O-])(=O)=O

Specifications

Color Yellow
Quantity 25 g
UN Number 1325
Formula Weight 270.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2,4-Dinitrobenzenesulfonate Hydrate
EINECSNumber (3)-2083
TSCA Yes