missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 1-Undecanesulfonate 98.0+%, TCI America™
Supplier: TCI America I034925G
Specifications
| Sodium 1-Undecanesulfonate [Reagent for Ion-Pair Chromatography] | |
| White | |
| MFCD00053627 | |
| sodium 1-undecanesulfonate, sodium undecane-1-sulfonate, 1-undecanesulfonic acid sodium salt, sodium undecylsulfonate, ipc-alks-11, acmc-1bn99, 1-undecanesulfonicacid, sodium salt 1:1 | |
| O=C(CN1C(=O)C2=CC=CC=C2C1=O)NC1=CC=C(C=C1)C(=O)NC1=NC=CS1 | |
| 406.42 | |
| 258.35 | |
| Crystalline Powder |
| 5838-34-6 | |
| C20H14N4O4S | |
| 25 g | |
| RFNALTSHRWGOHV-UHFFFAOYSA-N | |
| 4-[2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)acetamido]-N-(1,3-thiazol-2-yl)benzamide | |
| 23692564 | |
| ≥98.0% (T) |
Chemical Identifiers
| 5838-34-6 | |
| 406.42 | |
| RFNALTSHRWGOHV-UHFFFAOYSA-N | |
| 23692564 | |
| O=C(CN1C(=O)C2=CC=CC=C2C1=O)NC1=CC=C(C=C1)C(=O)NC1=NC=CS1 |
| C20H14N4O4S | |
| MFCD00053627 | |
| sodium 1-undecanesulfonate, sodium undecane-1-sulfonate, 1-undecanesulfonic acid sodium salt, sodium undecylsulfonate, ipc-alks-11, acmc-1bn99, 1-undecanesulfonicacid, sodium salt 1:1 | |
| 4-[2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)acetamido]-N-(1,3-thiazol-2-yl)benzamide |
Safety and Handling
EINECSNumber : (2)-1640
TSCA : No