missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Undecanesulfonate 98.0+%, TCI America™

Catalog No. p-7131356
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
I03495G 5 g
I034925G 25 g
2 options

Catalog No. I03495G

Supplier: TCI America I03495G

Only null left

Chemical Identifiers

CAS 5838-34-6
Molecular Formula C20H14N4O4S
Molecular Weight (g/mol) 406.42
MDL Number MFCD00053627
InChI Key RFNALTSHRWGOHV-UHFFFAOYSA-N
Synonym sodium 1-undecanesulfonate, sodium undecane-1-sulfonate, 1-undecanesulfonic acid sodium salt, sodium undecylsulfonate, ipc-alks-11, acmc-1bn99, 1-undecanesulfonicacid, sodium salt 1:1
PubChem CID 23692564
IUPAC Name 4-[2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)acetamido]-N-(1,3-thiazol-2-yl)benzamide
SMILES O=C(CN1C(=O)C2=CC=CC=C2C1=O)NC1=CC=C(C=C1)C(=O)NC1=NC=CS1

Specifications

Color White
Quantity 5 g
Formula Weight 258.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Undecanesulfonate [Reagent for Ion-Pair Chromatography]
EINECSNumber (2)-1640
TSCA No